Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2(3H)-Benzothiazolone 98.0+%, TCI America™
SDP

Catalog No. B234425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B234425G 25 g
B23445G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B234425G Supplier TCI America Supplier No. B234425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 934-34-9
Molecular Formula C7H5NOS
Molecular Weight (g/mol) 151.183
MDL Number MFCD00022868
InChI Key YEDUAINPPJYDJZ-UHFFFAOYSA-N
Synonym benzothiazolone, 2-hydroxybenzothiazole, 2 3h-benzothiazolone, 2-benzothiazolol, 2-benzothiazolone, 1,3-benzothiazol-2-ol, 2-benzothiazolinone, 3h-benzothiazol-2-one, benzothiazolol, benzo d thiazol-2 3h-one
PubChem CID 13625
ChEBI CHEBI:115196
IUPAC Name 3H-1,3-benzothiazol-2-one
SMILES C1=CC=C2C(=C1)NC(=O)S2

Specifications

Melting Point 138°C
Quantity 25 g
Formula Weight 151.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2(3H)-Benzothiazolone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.