Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,4-Trimethyl-2-pentene 97.0+%, TCI America™
SDP

Catalog No. T0780100ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0780100ML 100 mL
T078025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T0780100ML Supplier TCI America Supplier No. T0780100ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 107-40-4
Molecular Formula C8H16
Molecular Weight (g/mol) 112.216
MDL Number MFCD00008902
InChI Key LAAVYEUJEMRIGF-UHFFFAOYSA-N
Synonym 2,4,4-trimethyl-2-pentene, 2-pentene, 2,4,4-trimethyl, 2,2,4-trimethyl-3-pentene, 2,4,4-trimethylpentene-2, unii-548r0du21i, beta-diisobutylene, propene, 1-tert-butyl-2-methyl, acmc-1cujo, dsstox_cid_6766, dsstox_rid_78208
PubChem CID 7869
IUPAC Name 2,4,4-trimethylpent-2-ene
SMILES CC(=CC(C)(C)C)C

Specifications

Color Colorless
Boiling Point 105°C
Quantity 100 mL
UN Number 2050
Formula Weight 112.22
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,4,4-Trimethyl-2-pentene
EINECSNumber (2)-0024
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.