Learn More
Sigma Organic Chemistry 2 4 5 7-tetrabromo-3 4 | 1G | MFCD00067282
Supplier: Sigma Organic Chemistry CDS0032071G

2 4 5 7-tetrabromo-3 4 , 1G
About this Item:
MDL #: MFCD00067282
Molecular Weight: 785.67
UNSPSC Code: 12352200
Chemical Formula: C20H4Br4Cl4O5
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.