Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2,4,5-Trichlorophenol

Catalog No. 91332250GF Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
91-332-250GF 250 g
91-332-50GF 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 91-332-250GF Supplier Sigma Aldrich Supplier No. 91332250GF
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2,4,5-Trichlorophenol

Specifications

CAS 95-95-4
Molecular Formula C6H3Cl3O
Linear Formula Cl3C6H2OH
MDL Number MFCD00002170
Melting Point 64°C to 67°C, 67°C to 69°C (lit.)
Boiling Point 248°C (740 mmHg)
Molecular Weight (g/mol) 197.45
Quantity 250 g
Percent Purity ≥95.0% (GC)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.