Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,5-Triphenylimidazole 98.0+%, TCI America™
SDP

Catalog No. T099925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T099925G 25 g
T09995G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T099925G Supplier TCI America Supplier No. T099925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 484-47-9
Molecular Formula C21H16N2
Molecular Weight (g/mol) 296.373
MDL Number MFCD00005187
InChI Key RNIPJYFZGXJSDD-UHFFFAOYSA-N
Synonym 2,4,5-triphenylimidazole, lophine, lophin, 1h-imidazole, 2,4,5-triphenyl, 2,4,5-triphenyl-1,3-imidazole, unii-q6k46g80zd, imidazole, 2,4,5-triphenyl, chembl34173, 2,5-triphenylimidazole, imidazole,4,5-triphenyl
PubChem CID 10232
IUPAC Name 2,4,5-triphenyl-1H-imidazole
SMILES C1=CC=C(C=C1)C2=C(N=C(N2)C3=CC=CC=C3)C4=CC=CC=C4

Specifications

Melting Point 275°C
Color White-Yellow
Quantity 25 g
Formula Weight 296.37
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4,5-Triphenylimidazole
RTECSNumber NI8710000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.