Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,6-Tri-tert-butylaniline, 95%

Catalog No. AAL0136203
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0136203 1 g
AAL0136206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0136203 Supplier Thermo Scientific Chemicals Supplier No. L0136203
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 961-38-6 | C18H31N | 261.453 g/mol

2,4,6-Tri-tert-butylnitrobenzene (bulky amine) is used in the synthesis of monomeric iminophosphorane

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4,6-Tri-tert-butylnitrobenzene (bulky amine) is used in the synthesis of monomeric iminophosphorane

Solubility
Insoluble in water.

Notes
Light Sensitive. Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 961-38-6
Molecular Formula C18H31N
Molecular Weight (g/mol) 261.453
MDL Number MFCD00011645
InChI Key REJGDSCBQPJPQT-UHFFFAOYSA-N
Synonym 2,4,6-tri-tert-butylaniline, 2,4,6-tri-t-butylaniline, 2,4,6-tri tert-butyl aniline, aniline, tri-tert-butyl, 2,4,6-tri-tert-butylphenylamine, 2,4,6-tritert-butylphenylamine, benzenamine, 2,4,6-tris 1,1-dimethylethyl, aniline, 2,4,6-tri-tert-butyl, 2,4,6-tris tert-butyl phenylamine, pubchem16758
PubChem CID 70402
IUPAC Name 2,4,6-tritert-butylaniline
SMILES CC(C)(C)C1=CC(=C(C(=C1)C(C)(C)C)N)C(C)(C)C

Specifications

Melting Point 144°C to 147°C
Quantity 1 g
UN Number UN2811
Beilstein 2272612
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 261.46
Percent Purity 95%
Chemical Name or Material 2,4,6-Tri-tert-butylaniline
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 213-507-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.