Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,6-Tri-tert-butylnitrobenzene 95.0+%, TCI America™
SDP

Catalog No. T182125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T182125G 25 g
T18215G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T182125G Supplier TCI America Supplier No. T182125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4074-25-3
Molecular Formula C18H29NO2
Molecular Weight (g/mol) 291.435
MDL Number MFCD00008818
InChI Key IMDZOFHRUMJNQR-UHFFFAOYSA-N
Synonym 2,4,6-tri-tert-butylnitrobenzene, 2,4,6-tri-t-butylnitrobenzene, 1,3,5-tri-tert-butyl-2-nitrobenzene, 1,3,5-tri tert-butyl-2-nitrobenzene, 2,4,6 tri-tert-butylnitrobenzene, 2-nitro-1,3,5-tri-tert-butylbenzene, acmc-209jgh, 1,3,5-tritert-butyl-2-nitro-benzene, 2-nitro-1,3,5-tritert-butyl-benzene, 2,4,6-tri-tert-butyl-1-nitrobenzene
PubChem CID 77691
IUPAC Name 1,3,5-tritert-butyl-2-nitrobenzene
SMILES CC(C)(C)C1=CC(=C(C(=C1)C(C)(C)C)[N+](=O)[O-])C(C)(C)C

Specifications

Melting Point 206°C
Color Yellow
Quantity 25 g
Formula Weight 291.44
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4,6-Tri-tert-butylnitrobenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.