Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,6-Triamino-5-nitrosopyrimidine 98.0+%, TCI America™
SDP

Catalog No. T170525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T170525G 25 g
T17055G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T170525G Supplier TCI America Supplier No. T170525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1006-23-1
Molecular Formula C4H6N6O
Molecular Weight (g/mol) 154.13
MDL Number MFCD00006096
InChI Key XLQQJSWJHHKLOK-UHFFFAOYSA-N
PubChem CID 70504
IUPAC Name 5-nitrosopyrimidine-2,4,6-triamine
SMILES NC1=NC(N)=C(N=O)C(N)=N1

Specifications

Color Red
Quantity 25 g
Formula Weight 154.13
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4,6-Triamino-5-nitrosopyrimidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.