Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,6-Tribromo-3-hydroxybenzaldehyde, 98%, Thermo Scientific Chemicals

Catalog No. p-7044899
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0865609 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0865609 Supplier Thermo Scientific Chemicals Supplier No. L0865609
Only null left
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2737-22-6
Molecular Formula C7H3Br3O2
Molecular Weight (g/mol) 358.81
MDL Number MFCD00017320
InChI Key FAWOIFAFFUDNJX-UHFFFAOYSA-N
Synonym 3-hydroxy-2,4,6-tribromobenzaldehyde, benzaldehyde, 3-hydroxy-2,4,6-tribromo, benzaldehyde,2,4,6-tribromo-3-hydroxy, acmc-1cj0g, 3-08-00-00209 beilstein handbook reference, 2,4,6-tris bromanyl-3-oxidanyl-benzaldehyde
PubChem CID 200803
IUPAC Name 2,4,6-tribromo-3-hydroxybenzaldehyde
SMILES OC1=C(Br)C=C(Br)C(C=O)=C1Br

Specifications

Melting Point 119°C to 122°C
Quantity 10 g
Beilstein 2098037
Sensitivity Air Sensitive
Formula Weight 358.81
Percent Purity 98%
Chemical Name or Material 2,4,6-Tribromo-3-hydroxybenzaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
RTECSNumber CU6900000
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.