Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,6-Tribromopyridine 97.0+%, TCI America™
SDP

Catalog No. T33751G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T33751G 1 g
T33755G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T33751G Supplier TCI America Supplier No. T33751G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2408-70-0
Molecular Formula C5H2Br3N
Molecular Weight (g/mol) 315.79
MDL Number MFCD00234002
InChI Key WALXYTCBNHJWER-UHFFFAOYSA-N
PubChem CID 13495919
IUPAC Name 2,4,6-tribromopyridine
SMILES C1=C(C=C(N=C1Br)Br)Br

Specifications

Melting Point 109°C
Color White-Yellow
Boiling Point 284°C
Quantity 1 g
Formula Weight 315.79
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4,6-Tribromopyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.