Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,6-Trichloroaniline, 98+%

Catalog No. AAA1904414
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1904414 25 g
AAA1904422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1904414 Supplier Thermo Scientific Chemicals Supplier No. A1904414
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 634-93-5 | C6H4Cl3N | 196.455 g/mol

Tetrachloronitrobenzene was prepared from 2:4:6-trichloroaniline,which was converted by a Sandmeyer reaction into 1:2:3:5- tetrachlorobenzene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetrachloronitrobenzene was prepared from 2:4:6-trichloroaniline,which was converted by a Sandmeyer reaction into 1:2:3:5- tetrachlorobenzene.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 634-93-5
Molecular Formula C6H4Cl3N
Molecular Weight (g/mol) 196.455
MDL Number MFCD00007663
InChI Key NATVSFWWYVJTAZ-UHFFFAOYSA-N
Synonym s-trichloroaniline, benzenamine, 2,4,6-trichloro, aniline, 2,4,6-trichloro, sym-trichloroaniline, 2,4,6-trichlorobenzenamine, 2,4,6-trichlorophenylamine, 1-amino-2,4,6-trichlorobenzene, unii-j7ic72n9b0, ccris 601, pubchem9974
PubChem CID 12471
IUPAC Name 2,4,6-trichloroaniline
SMILES C1=C(C=C(C(=C1Cl)N)Cl)Cl

Specifications

Melting Point 75°C to 79°C
Quantity 25 g
UN Number UN2018
Beilstein 879568
Boiling Point 262°C
Solubility Information Insoluble in water.
Formula Weight 196.46
Percent Purity ≥98%
Chemical Name or Material 2,4,6-Trichloroaniline
Hazard Category H301+H311+H331-H373
Hazard Statement GHS H Statement
H301-H311-H331-H373
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement P260-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P314-P330-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: CHLOROANILINES, SOLID
EINECSNumber 211-219-8
RTECSNumber BZ0250000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.