Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,6-Trichlorobenzonitrile, 97+%

Catalog No. AAA1460906
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1460906 5 g
AAA1460903 1 g
AAA1460914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1460906 Supplier Thermo Scientific Chemicals Supplier No. A1460906
Add to Cart

CAS: 6575-05-9 | C7H2Cl3N | 206.45 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6575-05-9
MDL Number MFCD00052711

Specifications

Melting Point 80°C to 82°C
Quantity 5 g
Molecular Formula C7H2Cl3N
UN Number UN3439
Synonym benzonitrile, 2,4,6-trichloro, 2,4,6-trichlorobenzenecarbonitrile, pubchem4822, acmc-1b2tu, pgodhcioipodfe-uhfffaoysa
InChI Key PGODHCIOIPODFE-UHFFFAOYSA-N
SMILES C1=C(C=C(C(=C1Cl)C#N)Cl)Cl
IUPAC Name 2,4,6-trichlorobenzonitrile
Molecular Weight (g/mol) 206.45
PubChem CID 522179
Formula Weight 206.46
Percent Purity ≥97%
Chemical Name or Material 2,4,6-Trichlorobenzonitrile
Show More Show Less
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.