Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,6-Trichlorophenyl Isocyanate 98.0+%, TCI America™
SDP

Catalog No. I05651G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I05651G 1 g
I05655G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I05651G Supplier TCI America Supplier No. I05651G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2505-31-9
Molecular Formula C7H2Cl3NO
Molecular Weight (g/mol) 222.449
MDL Number MFCD00013845
InChI Key VDYWXVDWKFAUKE-UHFFFAOYSA-N
Synonym Isocyanic Acid 2,4,6-Trichlorophenyl Ester
PubChem CID 613574
IUPAC Name 1,3,5-trichloro-2-isocyanatobenzene
SMILES C1=C(C=C(C(=C1Cl)N=C=O)Cl)Cl

Specifications

Melting Point 67°C
Quantity 1 g
Physical Form Crystal-Powder at 20°C
UN Number 2206
Boiling Point 101°C
Formula Weight 222.45
Percent Purity ≥98.0% (GC)
Chemical Name or Material 2,4,6-Trichlorophenyl Isocyanate
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.