Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2',4',6'-Trifluoroacetophenone 98.0+%, TCI America™
SDP

Catalog No. T273325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T273325G 25 g
T27335G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T273325G Supplier TCI America Supplier No. T273325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 51788-77-3
Molecular Formula C8H5F3O
Molecular Weight (g/mol) 174.12
MDL Number MFCD00061194
InChI Key FFJSZHCBWFPSJI-UHFFFAOYSA-N
Synonym 2',4',6'-trifluoroacetophenone, 1-2,4,6-trifluorophenyl ethanone, 2,4,6-trifluoroacetophenone, 1-2,4,6-trifluorophenyl ethan-1-one, ethanone, 1-2,4,6-trifluorophenyl, 1-2,4,6-trifluoro-phenyl-ethanone, 1-acetyl-2,4,6-trifluorobenzene, pubchem4290, acmc-209kvx, 2',4,6'-trifluoroacetophenone
PubChem CID 2776906
IUPAC Name 1-(2,4,6-trifluorophenyl)ethan-1-one
SMILES CC(=O)C1=C(F)C=C(F)C=C1F

Specifications

Color Yellow
Boiling Point 190°C
Quantity 25 g
Formula Weight 174.12
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2′,4′,6′-Trifluoroacetophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.