Learn More
2,4,6-Triiodophenol, 98%
Description
2,4,6-Triiodophenol is used as an iodinated disinfection by product formed during chlorination of sewage effluents. It is also used as a potent thyroid disrupting chemical and to investigate deiodinase.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2,4,6-Triiodophenol is used as an iodinated disinfection by product formed during chlorination of sewage effluents. It is also used as a potent thyroid disrupting chemical and to investigate deiodinase.
Solubility
Insoluble in water.
Notes
Incompatible withacid chlorides, acid anhydrides and oxidizing agents.
Specifications
Specifications
| Melting Point | 156°C to 160°C |
| Quantity | 25 g |
| Beilstein | 2046861 |
| Sensitivity | Light sensitive |
| Solubility Information | Insoluble in water. |
| Formula Weight | 471.8 |
| Percent Purity | 98% |
| Chemical Name or Material | 2,4,6-Triiodophenol |
Safety and Handling
| Hazard Category | H312-H315-H319-H332-H335 |
| Hazard Statement | GHS H Statement H302-H315-H319-H335 Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| EINECSNumber | 210-186-7 |
| RTECSNumber | SN2800000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.