Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,6-Triisopropylbenzenesulfonyl Chloride 97%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
6553-96-4
C15H23ClO2S
302.85
MFCD00007433
JAPYIBBSTJFDAK-UHFFFAOYSA-N
2,4,6-triisopropylbenzenesulfonyl chloride,2,4,6-triisopropylbenzene-1-sulfonyl chloride,trisyl chloride,2,4,6-triisopropylbenzenesulphonyl chloride,benzenesulfonyl chloride, 2,4,6-tris 1-methylethyl,2,4,6-triisopropylbenzenesulfonylchloride,2,4,6-triisopropyl-benzenesulfonyl chloride,tpscl 2,4,6-triisopropyl benzenesulfonyl chloride,2,4,6-triisopropylphenylsulfonyl chloride,2,4,6-triisopropyl benzenesulfonyl chloride
81042
2,4,6-tri(propan-2-yl)benzenesulfonyl chloride
CC(C)C1=CC(=C(C(=C1)C(C)C)S(=O)(=O)Cl)C(C)C
6553-96-4
C15H23ClO2S
302.85
MFCD00007433
JAPYIBBSTJFDAK-UHFFFAOYSA-N
2,4,6-triisopropylbenzenesulfonyl chloride,2,4,6-triisopropylbenzene-1-sulfonyl chloride,trisyl chloride,2,4,6-triisopropylbenzenesulphonyl chloride,benzenesulfonyl chloride, 2,4,6-tris 1-methylethyl,2,4,6-triisopropylbenzenesulfonylchloride,2,4,6-triisopropyl-benzenesulfonyl chloride,tpscl 2,4,6-triisopropyl benzenesulfonyl chloride,2,4,6-triisopropylphenylsulfonyl chloride,2,4,6-triisopropyl benzenesulfonyl chloride
81042
2,4,6-tri(propan-2-yl)benzenesulfonyl chloride
CC(C)C1=CC(=C(C(=C1)C(C)C)S(=O)(=O)Cl)C(C)C
Packaging Glass bottle
Color White to Beige
Quantity 25g
Melting Point 93°C to 97°C
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula [(CH3)2CH]3C6H2SO2Cl
Fieser 01,1228; 03,308; 06,622
Solubility Information Solubility in water: decomposes
Formula Weight 302.85
Physical Form Crystalline Solid
Percent Purity ≥96.0%
Chemical Name or Material 2, 4, 6-Triisopropylbenzenesulfonyl chloride, 97%
Show More Show Less

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 229-479-6

TSCA : TSCA