Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,6-Trimethoxypyrimidine 98.0+%, TCI America™
SDP

Catalog No. T20395G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T20395G 5 g
T20391G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T20395G Supplier TCI America Supplier No. T20395G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 13106-85-9
Molecular Formula C7H10N2O3
Molecular Weight (g/mol) 170.168
MDL Number MFCD00829225
InChI Key RJVAFLZWVUIBOU-UHFFFAOYSA-N
Synonym acmc-209bkb, 2,4,6-trimethoxy-pyrimidine, pyrimidine,2,4,6-trimethoxy, rjvaflzwvuibou-uhfffaoysa, 2,4,6-trimethoxypyrimidine
PubChem CID 726938
IUPAC Name 2,4,6-trimethoxypyrimidine
SMILES COC1=CC(=NC(=N1)OC)OC

Specifications

Melting Point 54°C
Color White
Boiling Point 232°C
Quantity 5 g
Formula Weight 170.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4,6-Trimethoxypyrimidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.