Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,6-Trimethylbenzyl Alcohol 98.0+%, TCI America™
SDP

Catalog No. T190525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T190525G 25 g
T19055G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T190525G Supplier TCI America Supplier No. T190525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4170-90-5
Molecular Formula C10H14O
Molecular Weight (g/mol) 150.221
MDL Number MFCD00014422
InChI Key LODDFDHPSIYCTK-UHFFFAOYSA-N
Synonym 2,4,6-trimethylbenzyl alcohol, mesitylmethanol, mesitylcarbinol, 2,4,6-trimethylphenyl methanol, unii-rev3kvt84n, benzenemethanol, 2,4,6-trimethyl, 2,4,6-trimethylbenzylic alcohol, benzyl alcohol, 2,4,6-trimethyl, rev3kvt84n, 2,4,6-trimethylphenyl methan-1-ol
PubChem CID 20139
IUPAC Name (2,4,6-trimethylphenyl)methanol
SMILES CC1=CC(=C(C(=C1)C)CO)C

Specifications

Melting Point 88°C
Color White
Boiling Point 141°C
Quantity 25 g
Formula Weight 150.22
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4,6-Trimethylbenzyl Alcohol
RTECSNumber DP1402000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.