Learn More
2,4,6-Trimethylstyrene, 95%, stab. with 500ppm 4-tert-butylcatechol
Description
2,4,6-Trimethylstyrene is used as an intermediate to produce other chemicals like 2-Methoxy-1-(2, 4, 6-trimethyl-phenyl)-ethanone.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2,4,6-Trimethylstyrene is used as an intermediate to produce other chemicals like 2-Methoxy-1-(2, 4, 6-trimethyl-phenyl)-ethanone.
Solubility
Soluble in methanol.
Notes
It is sensitive to light. Incompatible with oxidizing agents.
Specifications
Specifications
| Density | 0.908 |
| Boiling Point | 208°C to 209°C |
| Flash Point | 75°C (167°F) |
| Refractive Index | 1.5295 |
| Quantity | 5 g |
| Beilstein | 1904365 |
| Solubility Information | Soluble in methanol. |
| Formula Weight | 146.23 |
| Percent Purity | 95% |
| Chemical Name or Material | 2,4,6-Trimethylstyrene, stabilized with 500ppm 4-tert-butylcatechol |
Safety and Handling
| Hazard Category | Warning |
| Hazard Statement | GHS H Statement H315-H319-H335-H227 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. Combustible liquid. |
| Precautionary Statement | GHS P Statement P210-P261-P280-P305+P351+P338-P405-P501a Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Avoid breathing dust/fume/gas/mist/vapors/spray. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Store locked up. Dispose of contents/container in accordance with local/regional/national/international regulations. |
| EINECSNumber | 212-205-4 |
| TSCA | No |
| Recommended Storage | Keep cold |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.