Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,6-Tris(3-bromophenyl)-1,3,5-triazine 98.0+%, TCI America™
SDP

Catalog No. T33081G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T33081G 1 g
T33085G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T33081G Supplier TCI America Supplier No. T33081G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 890148-78-4
Molecular Formula C21H12Br3N3
Molecular Weight (g/mol) 546.06
InChI Key IWHHYACTSSPXDV-UHFFFAOYSA-N
PubChem CID 57872377
IUPAC Name 2,4,6-tris(3-bromophenyl)-1,3,5-triazine
SMILES C1=CC(=CC(=C1)Br)C2=NC(=NC(=N2)C3=CC(=CC=C3)Br)C4=CC(=CC=C4)Br

Specifications

Melting Point 209°C
Color White-Yellow
Quantity 1 g
Formula Weight 546.06
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 2,4,6-Tris(3-bromophenyl)-1,3,5-triazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.