Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,6-Tris(dimethylaminomethyl)phenol, Thermo Scientific™

Catalog No. AC469290250
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC469290250 25 g
AC469291000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AC469290250 Supplier Thermo Scientific Chemicals Supplier No. 469290250
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Alpha Vector TRISDIMETHYLAMINOMETHYLPHENOL
Chemical Name or Material 2,4,6-Tris(dimethylaminomethyl)phenol
CAS 90-72-2
Color Colorless to Yellow
Flash Point 149°C
Infrared Spectrum Conforms
Packaging Glass bottle
Molecular Formula C15 H27 N3 O
Refractive Index 1.5150 to 1.5170 (20°C, 589 nm)
MDL Number 00008330
Quantity 25 g
UN Number 2735
Molecular Weight (g/mol) 265.4
Physical Form Viscous Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.