Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam 2-(4-Aminophenyl)-6-Methylbenzothiazole-7-Sulphonic Acid, Fluorescent Labeling Reagent, Solid, >95% Purity, 10 g

Catalog No. 502665745
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-266-5745 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-266-5745 Supplier Abcam Supplier No. AB14551210G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

High-purity fluorescent labeling reagent with >95% purity.

  • Molecular weight: 320.39 g/mol
  • Purity: >95%
  • Form: Solid
  • Components: 100% 2-(4-aminophenyl)-6-methylbenzothiazole-7-sulphonic acid
  • CAS number: 130-17-6
  • Storage: Store at +4°C

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Substrate Properties Chemical Substrate
Shipping Condition Ambient
Label or Dye Fluorescent Labeling Reagent
Product Type Protein Labeling Reagent
Form Solid
Buffer 100% 2-(4-aminophenyl)-6-methylbenzothiazole-7-sulphonic acid
Content And Storage Store at 4°C.
Detection Method Fluorescence
Formulation 100% 2-(4-aminophenyl)-6-methylbenzothiazole-7-sulphonic acid
Quantity 10 g
For Use With (Application) Fluorescent labeling
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.