Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(4-Bromophenyl)-5-fluoroindole, 97%, Thermo Scientific™

Catalog No. AAH6309103
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6309103 1 g
AAH63091MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6309103 Supplier Thermo Scientific Chemicals Supplier No. H6309103

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents, heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 885266-74-0
Molecular Formula C14H9BrFN
Molecular Weight (g/mol) 290.135
MDL Number MFCD05224214
InChI Key XAKLXLIHCVKNII-UHFFFAOYSA-N
Synonym 2-4-bromophenyl-5-fluoroindole, 2-4-bromophenyl-5-fluoro-1h-indole
PubChem CID 24720950
IUPAC Name 2-(4-bromophenyl)-5-fluoro-1H-indole
SMILES C1=CC(=CC=C1C2=CC3=C(N2)C=CC(=C3)F)Br

Specifications

Quantity 1 g
Formula Weight 290.13
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2-(4-Bromophenyl)-5-fluoroindole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.