Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(4-Chloro-3-nitrobenzoyl)benzoic Acid 97.0+%, TCI America™
SDP

Catalog No. C1013250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C1013250G 250 g
C101325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C1013250G Supplier TCI America Supplier No. C1013250G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 85-54-1
Molecular Formula C14H8ClNO5
Molecular Weight (g/mol) 305.67
MDL Number MFCD00007082
InChI Key RITAQDHCJBLSSL-UHFFFAOYSA-N
Synonym 2-4-chloro-3-nitrobenzoyl benzoic acid, benzoic acid, 2-4-chloro-3-nitrobenzoyl, 3'-nitro-4'-chlorobenzoylbenzoic acid, benzoic acid, o-4-chloro-3-nitrobenzoyl, 2-4-chloro-3-nitrophenyl carbonyl benzoic acid, acmc-209q6m, ksc496o4t, o-4-chloro-3-nitrobenzoyl benzoic acid, 2-4-chloro-3-nitro-benzoyl benzoic acid, 2-4'-chloro-3'-nitrobenzoyl benzoic acid
PubChem CID 66562
IUPAC Name 2-(4-chloro-3-nitrobenzoyl)benzoic acid
SMILES OC(=O)C1=CC=CC=C1C(=O)C1=CC=C(Cl)C(=C1)[N+]([O-])=O

Specifications

Melting Point 200°C
Color White-Yellow
Quantity 250 g
Formula Weight 305.67
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(4-Chloro-3-nitrobenzoyl)benzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.