Learn More
Sigma Organic Chemistry 2- 4-Chlorophenoxy -1- | 1G | 115781-11-8 | MFCD00425148
Supplier: Sigma Organic Chemistry OTV0001591G

2- 4-Chlorophenoxy -1- , 1G
About this item:
CAS #: 115781-11-8
MDL #: MFCD00425148
Chemical Formula: C14H11ClO4
Molecular Weight: 278.69
UNSPSC Code: 12352200
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.