Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(4-Chlorophenyl)ethyl isothiocyanate, 97%, Thermo Scientific™

Catalog No. AAA1548703
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1548703 1 g
AAA1548706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1548703 Supplier Thermo Scientific Chemicals Supplier No. A1548703

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17608-10-5
Molecular Formula C9H8ClNS
Molecular Weight (g/mol) 197.68
MDL Number MFCD00041127
InChI Key MRJJYUJULSZFDV-UHFFFAOYSA-N
Synonym 2-4-chlorophenyl ethyl isothiocyanate, 1-chloro-4-2-isothiocyanatoethyl benzene, 2-4-chlorophenethyl isothiocyanate, 2-4-chlorophenethyl isothicyanate, acmc-1bsas, 2-4-chlorophenyl ethanisothiocyanate, 1-chloranyl-4-2-isothiocyanatoethyl benzene, 1-chloro-4-2-isothiocyanatoethyl benzene #
PubChem CID 140257
IUPAC Name 1-chloro-4-(2-isothiocyanatoethyl)benzene
SMILES C1=CC(=CC=C1CCN=C=S)Cl

Specifications

Quantity 1 g
UN Number UN2922
Beilstein 2719060
Boiling Point 145°C (1 mmHg)
Formula Weight 197.69
Percent Purity 97%
Chemical Name or Material 2-(4-Chlorophenyl)ethyl isothiocyanate
Hazard Category Danger
Hazard Statement GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.