Learn More
Sigma Organic Chemistry 2- 4-chlorophenyl indol | 1G | 1211-35-4 | MFCD00047159
Supplier: Sigma Organic Chemistry CDS0135521G

2- 4-chlorophenyl indol , 1G
About this Item:
CAS #: 1211-35-4
MDL #: MFCD00047159
Molecular Weight: 227.69
UNSPSC Code: 12352200
Chemical Formula: C14H10ClN
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.