Learn More
Sigma Organic Chemistry 2- 4-Cyanobenzylthio ac 1G
Supplier: Sigma Organic Chemistry OTV0007861G

2- 4-Cyanobenzylthio ac , 1G
About this item:
MDL #: MFCD01030701
Chemical Formula: C10H9NO2S
Molecular Weight: 207.25
UNSPSC Code: 12352200
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.