Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-D methyl ester, Restek
SDP

Catalog No. 06719866 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-719-866 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-719-866 Supplier Restek Corporation Supplier No. 32240
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1928-38-7
Molecular Formula C9H8Cl2O3
Molecular Weight (g/mol) 235.06
InChI Key HWIGZMADSFQMOI-UHFFFAOYSA-N
PubChem CID 1488
IUPAC Name methyl 2-(2,4-dichlorophenoxy)acetate
SMILES COC(=O)COC1=C(C=C(C=C1)Cl)Cl

Specifications

Quantity 1 mL
Chemical Name or Material 2, 4-D methyl ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.