Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Diamino-1,3,5-triazine 98.0+%, TCI America™
SDP

Catalog No. D22271G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D22271G 1 g
D22275G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D22271G Supplier TCI America Supplier No. D22271G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 504-08-5
Molecular Formula C3H5N5
Molecular Weight (g/mol) 111.108
MDL Number MFCD00014598
InChI Key VZXTWGWHSMCWGA-UHFFFAOYSA-N
PubChem CID 10435
ChEBI CHEBI:38071
IUPAC Name 1,3,5-triazine-2,4-diamine
SMILES C1=NC(=NC(=N1)N)N

Specifications

Melting Point 325°C
Color White-Yellow
Quantity 1 g
Formula Weight 111.11
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Diamino-1,3,5-triazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.