Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Diamino-6-chloropyrimidine, 98%, Thermo Scientific™

Catalog No. AAA1778006
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1778006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1778006 Supplier Thermo Scientific Chemicals Supplier No. A1778006

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 156-83-2
Molecular Formula C4H5ClN4
Molecular Weight (g/mol) 144.562
MDL Number MFCD00006097
InChI Key QJIUMVUZDYPQRT-UHFFFAOYSA-N
Synonym 4-chloro-2,6-diaminopyrimidine, 2,4-diamino-6-chloropyrimidine, 2,6-diamino-4-chloropyrimidine, 6-chloro-2,4-diaminopyrimidine, 6-chloro-pyrimidine-2,4-diamine, 2,4-pyrimidinediamine, 6-chloro, 4-chloro-2,6-diamino pyrimidine, 2-amino-6-chloro-4-pyrimidinylamine, caap, minoxidil impurity b
PubChem CID 67432
IUPAC Name 6-chloropyrimidine-2,4-diamine
SMILES C1=C(N=C(N=C1Cl)N)N

Specifications

Melting Point 199°C to 202°C
Quantity 5 g
Beilstein 118455
Formula Weight 144.56
Percent Purity 98%
Chemical Name or Material 2,4-Diamino-6-chloropyrimidine
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 205-863-9
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.