Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dichloro-3,5-dimethylphenol 98.0+%, TCI America™
SDP

Catalog No. D110925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D110925G 25 g
D1109500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D110925G Supplier TCI America Supplier No. D110925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 133-53-9
Molecular Formula C8H8Cl2O
Molecular Weight (g/mol) 191.051
MDL Number MFCD00019981
InChI Key IYOLBFFHPZOQGW-UHFFFAOYSA-N
Synonym dichloroxylenol, dcmx, ottacide, decasept, hewsol, prinsyl, dixol, 2,4-dichloro-3,5-xylenol, 3,5-dimethyl-2,4-dichlorophenol, unii-51ac49olt7
PubChem CID 8621
IUPAC Name 2,4-dichloro-3,5-dimethylphenol
SMILES CC1=CC(=C(C(=C1Cl)C)Cl)O

Specifications

Melting Point 93°C
Color Red-White
Quantity 25 g
UN Number 2430
Formula Weight 191.05
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Dichloro-3,5-dimethylphenol
EINECSNumber (3)-0989
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.