Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dichloro-5-methylpyrimidine 98.0+%, TCI America™
SDP

Catalog No. D361325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D361325G 25 g
D36131G 1 g
D36135G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D361325G Supplier TCI America Supplier No. D361325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1780-31-0
Molecular Formula C5H4Cl2N2
Molecular Weight (g/mol) 163.001
MDL Number MFCD00023197
InChI Key DQXNTSXKIUZJJS-UHFFFAOYSA-N
Synonym pyrimidine, 2,4-dichloro-5-methyl, 5-methyl-2,4-dichloropyrimidine, 2,4-dichloro-5-methyl-pyrimidine, 2,4-dichloro-5-methyl pyrimidine, 2, 4-dichloro-5-methylpyrimidine, 2,4-dichlor-5-methylpyrimidin, pubchem5288, zlchem 1254, acmc-1bsov, ksc178k6p
PubChem CID 74508
IUPAC Name 2,4-dichloro-5-methylpyrimidine
SMILES CC1=CN=C(N=C1Cl)Cl

Specifications

Color White-Yellow
Boiling Point 109°C
Quantity 25 g
Formula Weight 163.00
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2,4-Dichloro-5-methylpyrimidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.