Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dichloro-6-methylpyridine, 97%, Thermo Scientific™

Catalog No. AC453770050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC453770050 5 g
AC453770010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC453770050 Supplier Thermo Scientific Chemicals Supplier No. 453770050

Chemical Identifiers

CAS 42779-56-6
Molecular Formula C6H5Cl2N
Molecular Weight (g/mol) 162.02
InChI Key WUGTXQVNSRFDNV-UHFFFAOYSA-N
Synonym 2,4-dichloro-6-picoline, 2,4-dichloro-6-methyl-pyridine, pubchem5465, d0ri6l, 2-methyl-4,6-dichloropyridine, pyridine,2,4-dichloro-6-methyl
PubChem CID 11959083
IUPAC Name 2,4-dichloro-6-methylpyridine
SMILES CC1=NC(=CC(=C1)Cl)Cl

Specifications

Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Quantity 5 g
Formula Weight 162.02
Percent Purity 97%
Chemical Name or Material 2,4-Dichloro-6-methylpyridine
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. Harmful if inhaled. May cause respiratory irritation.
Precautionary Statement GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.