Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dichloro-6-(trifluoromethyl)pyrimidine, 95%, Thermo Scientific™

Catalog No. AC453792500
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC453792500 250 mg
AC453790010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC453792500 Supplier Thermo Scientific Chemicals Supplier No. 453792500

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 16097-64-6
Molecular Formula C5HCl2F3N2
Molecular Weight (g/mol) 216.98
InChI Key ZTNFYAJHLPMNSN-UHFFFAOYSA-N
Synonym 2,4-dichloro-6-trifluoromethyl pyrimidine, pyrimidine, 2,4-dichloro-6-trifluoromethyl, 2,4-dichloro-6-trifluoromethyl-pyrimidine, pubchem13247, ksc496a7p, 2,6-dichloro-4-trifluoromethyl pyrimidine
PubChem CID 334203
IUPAC Name 2,4-dichloro-6-(trifluoromethyl)pyrimidine
SMILES C1=C(N=C(N=C1Cl)Cl)C(F)(F)F

Specifications

Boiling Point 207°C
Flash Point 78°C
Assay Percent Range 94% min. (GC)
Quantity 250 mg
Formula Weight 216.98
Percent Purity 95%
Chemical Name or Material 2, 4-Dichloro-6-(trifluoromethyl)pyrimidine
Hazard Category GHS Signal Word: Warning

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.