Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2',4'-Dichloroacetophenone, 94%, Thermo Scientific™

Catalog No. AC176381000
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC176381000 100 g
AC176385000 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC176381000 Supplier Thermo Scientific Chemicals Supplier No. 176381000

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2234-16-4
Molecular Formula C8H6Cl2O
Molecular Weight (g/mol) 189.035
InChI Key XMCRWEBERCXJCH-UHFFFAOYSA-N
Synonym 2',4'-dichloroacetophenone, 1-2,4-dichlorophenyl ethanone, 2,4-dichloroacetophenone, ethanone, 1-2,4-dichlorophenyl, 1-2,4-dichlorophenyl ethan-1-one, p-chloro-2-chloroacetophenone, acetophenone, 2',4'-dichloro, unii-t7f2608h5h, 2',4'-dichloro phenyl ethanone, 1-2,4-dichloro-phenyl-ethanone
PubChem CID 16693
IUPAC Name 1-(2,4-dichlorophenyl)ethanone
SMILES CC(=O)C1=C(C=C(C=C1)Cl)Cl

Specifications

CAS Min % 92.0
CAS Max % 100.0
Density 1.32g/mL
Assay Percent Range 94%
Linear Formula Cl2C6H3COCH3
Quantity 100 g
Beilstein 07, II, 219
Formula Weight 189.04
Chemical Name or Material 2', 4'-Dichloroacetophenone, 94%
Hazard Statement GHS H Statement:
Harmful if swallowed.
Harmful to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement:
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 218-780-8
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.