Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dichlorobenzophenone 97.0+%, TCI America™
SDP

Catalog No. D034225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D034225G 25 g
D03425G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D034225G Supplier TCI America Supplier No. D034225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 19811-05-3
Molecular Formula C13H8Cl2O
Molecular Weight (g/mol) 251.11
MDL Number MFCD00018374
InChI Key VLTYTTRXESKBKI-UHFFFAOYSA-N
PubChem CID 72867
IUPAC Name (2,4-dichlorophenyl)(phenyl)methanone
SMILES ClC1=CC=C(C(=O)C2=CC=CC=C2)C(Cl)=C1

Specifications

Melting Point 47°C
Color White
Quantity 25 g
Formula Weight 251.11
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Dichlorobenzophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.