Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dichloronicotinic Acid 98.0+%, TCI America™
SDP

Catalog No. D49011G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D49011G 1 g
D49015G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D49011G Supplier TCI America Supplier No. D49011G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 262423-77-8
Molecular Formula C6H3Cl2NO2
Molecular Weight (g/mol) 192.00
MDL Number MFCD06796292
InChI Key ZFGZNVSNOJPGDV-UHFFFAOYSA-N
Synonym 2,4-dichloronicotinic acid, 2,4-dichloronicotinc acid, 2,4-dichloronicotinicacid, 2,4-dichloro-3-pyridinecarboxylic acid, 2.4-dichloronicotinic acid, 2,4-dichloropyridine-3-carboxylicacid, dichloronicotinic acid;, pubchem17746, 2,4-dichloronicotinic acid;
PubChem CID 10583887
IUPAC Name 2,4-dichloropyridine-3-carboxylic acid
SMILES OC(=O)C1=C(Cl)C=CN=C1Cl

Specifications

Melting Point 151°C
Color White
Quantity 1 g
Formula Weight 192.00
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Dichloronicotinic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.