Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dichlorophenacyl Bromide 98.0+%, TCI America™
SDP

Catalog No. D335125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D335125G 25 g
D33515G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D335125G Supplier TCI America Supplier No. D335125G

Chemical Identifiers

CAS 2631-72-3
Molecular Formula C8H5BrCl2O
Molecular Weight (g/mol) 267.931
MDL Number MFCD00053005
InChI Key DASJDMQCPIDJIF-UHFFFAOYSA-N
Synonym 2-bromo-2',4'-dichloroacetophenone, 2-bromo-1-2,4-dichlorophenyl ethanone, 2,4-dichlorophenacyl bromide, 2-bromo-1-2,4-dichlorophenyl ethan-1-one, 2-bromo-1-2,4-dichloro-phenyl-ethanone, ethanone, 2-bromo-1-2,4-dichlorophenyl, 2'-bromo-2,4-dichloro acetophenone, omega-bromo-2,4-dichloro acetophenone, 2',4'-dichlorophenacyl bromide, 2-bromo-2,4-dichloroacetophenone
PubChem CID 75828
IUPAC Name 2-bromo-1-(2,4-dichlorophenyl)ethanone
SMILES C1=CC(=C(C=C1Cl)Cl)C(=O)CBr

Specifications

Color White
Quantity 25 g
UN Number 3261
Formula Weight 267.93
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2,4-Dichlorophenacyl Bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.