Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dichlorophenoxyacetic acid, 98%

Catalog No. AAA1246718
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1246718 50 g
AAA1246730 250 g
AAA124670B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1246718 Supplier Thermo Scientific Chemicals Supplier No. A1246718
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 94-75-7 | C8H6Cl2O3 | 221.033 g/mol

2,4-Dichlorophenoxyacetic acid is used as a supplement in plant cell culture media. It is a synthetic auxin, which finds application as a plant growth regulator. Further, it is used to increase latex output of old rubber trees.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Dichlorophenoxyacetic acid is used as a supplement in plant cell culture media. It is a synthetic auxin, which finds application as a plant growth regulator. Further, it is used to increase latex output of old rubber trees.

Solubility
Slightly soluble in water.

Notes
Moisture and light-sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94-75-7
Molecular Formula C8H6Cl2O3
Molecular Weight (g/mol) 221.033
MDL Number MFCD00004300
InChI Key OVSKIKFHRZPJSS-UHFFFAOYSA-N
Synonym 2,4-dichlorophenoxyacetic acid, 2-2,4-dichlorophenoxy acetic acid, 2,4-dichlorophenoxy acetic acid, hedonal, rhodia, agrotect, fernesta, fernimine, netagrone, tributon
PubChem CID 1486
ChEBI CHEBI:28854
IUPAC Name 2-(2,4-dichlorophenoxy)acetic acid
SMILES C1=CC(=C(C=C1Cl)Cl)OCC(=O)O

Specifications

Melting Point 136°C to 140°C
Density 1.42
Boiling Point 160°C (0.4 mmHg)
Odor Odorless
Quantity 50 g
Beilstein 1214242
Merck Index 14,2796
Solubility Information Slightly soluble in water.
Formula Weight 221.04
Percent Purity 98%
Chemical Name or Material 2,4-Dichlorophenoxyacetic acid
Show More Show Less
Hazard Category H302-H317-H318-H335-H351
Hazard Statement GHS H Statement
H318-H302-H312-H317-H335
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
May cause an allergic skin reaction.
May cause respiratory irritation.
Precautionary Statement P201-P202-P261-P264b-P270-P271-P272-P280g-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P310-P330-P333+P313-P363-P501c
EINECSNumber 202-361-1
RTECSNumber AG6825000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.