Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dichlorophenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. D24945G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D2494-5G 5 g
D2494-25G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D2494-5G Supplier TCI America Supplier No. D24945G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 68716-47-2
Molecular Formula C6H5BCl2O2
Molecular Weight (g/mol) 190.81
MDL Number MFCD00013930
InChI Key QNEGDGPAXKYZHZ-UHFFFAOYSA-N
Synonym 2,4-dichlorophenyl boronic acid, 2,4-dichlorobenzeneboronic acid, 2,4-dichlorophenylbornic acid, 2,4-dichlorophenyl boranediol, chembl20726
PubChem CID 2734659
IUPAC Name (2,4-dichlorophenyl)boronic acid
SMILES OB(O)C1=CC=C(Cl)C=C1Cl

Specifications

Melting Point 249°C
Color White-Yellow
Quantity 5 g
Formula Weight 190.81
Physical Form Crystalline Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.