Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dichlorophenylhydrazine Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. D2433250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D2433250G 250 g
D24335G 5 g
D243325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D2433250G Supplier TCI America Supplier No. D2433250G

Chemical Identifiers

CAS 5446-18-4
Molecular Formula C6H7Cl3N2
Molecular Weight (g/mol) 213.486
MDL Number MFCD00012929
InChI Key DDWYGJVFURAIJZ-UHFFFAOYSA-N
Synonym 2,4-dichlorophenylhydrazine hydrochloride, 2,4-dichlorophenyl hydrazine hydrochloride, 2,4-dichlorophenyl hydrazine monohydrochloride, 2,4-dichlorophenylhydrazine hcl, 2,4-dichloro-phenyl-hydrazine hydrochloride, 1-2,4-dichlorophenyl hydrazine hydrochloride, hydrazine, 2,4-dichlorophenyl-, monohydrochloride, pubchem7524, acmc-209lgs, ksc491e2h
PubChem CID 2723905
IUPAC Name (2,4-dichlorophenyl)hydrazine;hydrochloride
SMILES C1=CC(=C(C=C1Cl)Cl)NN.Cl

Specifications

Quantity 250 g
Formula Weight 213.49
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Dichlorophenylhydrazine Hydrochloride
EINECSNumber (3)-0472
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.