Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dichloroquinoline 98.0+%, TCI America™
SDP

Catalog No. D445225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D445225G 25 g
D44525G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D445225G Supplier TCI America Supplier No. D445225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 703-61-7
Molecular Formula C9H5Cl2N
Molecular Weight (g/mol) 198.05
MDL Number MFCD00023939
InChI Key QNBJYUUUYZVIJP-UHFFFAOYSA-N
PubChem CID 607503
IUPAC Name 2,4-dichloroquinoline
SMILES ClC1=CC(Cl)=C2C=CC=CC2=N1

Specifications

Melting Point 65°C
Boiling Point 282°C
Quantity 25 g
Formula Weight 198.05
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Dichloroquinoline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.