Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dichlorothiazole 98.0+%, TCI America™
SDP

Catalog No. D354925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D354925G 25 g
D35495G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D354925G Supplier TCI America Supplier No. D354925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4175-76-2
Molecular Formula C3HCl2NS
Molecular Weight (g/mol) 154.008
MDL Number MFCD08691358
InChI Key ICETWLGKJXCIDX-UHFFFAOYSA-N
PubChem CID 12333155
IUPAC Name 2,4-dichloro-1,3-thiazole
SMILES C1=C(N=C(S1)Cl)Cl

Specifications

Melting Point 44°C
Boiling Point 184°C
Quantity 25 g
Formula Weight 154.01
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Dichlorothiazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.