Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Diethylthioxanthen-9-one 98.0+%, TCI America™
SDP

Catalog No. D2375250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D2375250G 250 g
D237525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. D2375250G Supplier TCI America Supplier No. D2375250G
Add to Cart
Add to Cart
This item is not returnable. View return policy

Chemical Identifiers

CAS 82799-44-8
Molecular Formula C17H16OS
Molecular Weight (g/mol) 268.374
MDL Number MFCD00082910
InChI Key BTJPUDCSZVCXFQ-UHFFFAOYSA-N
PubChem CID 122825
IUPAC Name 2,4-diethylthioxanthen-9-one
SMILES CCC1=CC(=C2C(=C1)C(=O)C3=CC=CC=C3S2)CC

Specifications

Melting Point 70°C
Color Yellow
Quantity 250 g
Formula Weight 268.37
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Diethylthioxanthen-9-one
EINECSNumber (5)-5645
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.