Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Difluoro-3-iodopyridine, 95%, Thermo Scientific™

Catalog No. AAH6628003
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6628003 1 g
AAH66280MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6628003 Supplier Thermo Scientific Chemicals Supplier No. H6628003

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 837364-88-2
Molecular Formula C5H2F2IN
Molecular Weight (g/mol) 240.979
MDL Number MFCD09029861
InChI Key AYKASHJPLZSRDC-UHFFFAOYSA-N
Synonym 2,4-difluoro-3-iodo-pyridine, pyridine, 2,4-difluoro-3-iodo, 2,4-difluoroi-3-iodo-pyridine
PubChem CID 12175804
IUPAC Name 2,4-difluoro-3-iodopyridine
SMILES C1=CN=C(C(=C1F)I)F

Specifications

Quantity 1 g
Beilstein 9995192
Sensitivity Light sensitive
Formula Weight 240.98
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 2,4-Difluoro-3-iodopyridine
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.