Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Difluorobenzenesulfonamide 98.0+%, TCI America™
SDP

Catalog No. D516125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D516125G 25 g
D51615G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D516125G Supplier TCI America Supplier No. D516125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 13656-60-5
Molecular Formula C6H5F2NO2S
Molecular Weight (g/mol) 193.168
MDL Number MFCD01320743
InChI Key PTHLPYKPCQLAOF-UHFFFAOYSA-N
PubChem CID 574964
IUPAC Name 2,4-difluorobenzenesulfonamide
SMILES C1=CC(=C(C=C1F)F)S(=O)(=O)N

Specifications

Melting Point 156°C
Color White
Quantity 25 g
Synonym 2,4-difluorobenzene-1-sulfonamide, 2,4-difluorobenzenesulphonamide, benzenesulfonamide, 2,4-difluoro, benzenesulfonamide,2,4-difluoro, buttpark 23\07-80, 2,4-difluoro-1-benzenesulfonamide, 2,4-difluorobenzenesulfoneamide, pubchem11764, enamine_001177, acmc-1c82s
Formula Weight 193.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Difluorobenzenesulfonamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.