Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4'-Difluorobenzophenone 96.0+%, TCI America™
SDP

Catalog No. D179625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D179625G 25 g
D17965G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D179625G Supplier TCI America Supplier No. D179625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 342-25-6
Molecular Formula C13H8F2O
Molecular Weight (g/mol) 218.203
MDL Number MFCD00000319
InChI Key LKFIWRPOVFNPKR-UHFFFAOYSA-N
Synonym 2,4'-difluorobenzophenone, 2-fluorophenyl 4-fluorophenyl methanone, 2',4-difluorobenzophenone, 2-fluorophenyl-4-fluorophenyl methanone, di2-fluorophenyl ketone, fr bvr df, pubchem7479, acmc-1clee, 2,4;-difluorobenzophenone, 2,4/'-difluorobenzophenone
PubChem CID 67651
IUPAC Name (2-fluorophenyl)-(4-fluorophenyl)methanone
SMILES C1=CC=C(C(=C1)C(=O)C2=CC=C(C=C2)F)F

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 218.20
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2,4′-Difluorobenzophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.