Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Difluorophenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. D339125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D3391-25G 25 g
D33915G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D3391-25G Supplier TCI America Supplier No. D339125G
Add to Cart
Add to Cart

Specifications

CAS 144025-03-6
Melting Point 250°C
Color White-Yellow
Molecular Formula C6H5BF2O2
Quantity 25 g
MDL Number MFCD01318998
Synonym 2,4-difluorobenzeneboronic acid, 2,4-difluorophenyl boronic acid, 2,4-fluorobenzene boronic acid, 2,4-difluorophenyl boranediol, 2,4-difluorophenyl-boronic acid
InChI Key QQLRSCZSKQTFGY-UHFFFAOYSA-N
SMILES B(C1=C(C=C(C=C1)F)F)(O)O
IUPAC Name (2,4-difluorophenyl)boronic acid
Molecular Weight (g/mol) 157.911
PubChem CID 2734334
Formula Weight 157.91
Physical Form Crystalline Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.