Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2',4'-Difluoropropiophenone 98.0+%, TCI America™
SDP

Catalog No. D259825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D259825G 25 g
D25985G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D259825G Supplier TCI America Supplier No. D259825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 85068-30-0
Molecular Formula C9H8F2O
Molecular Weight (g/mol) 170.16
MDL Number MFCD00015507
InChI Key UZWOADNMVRRYDE-UHFFFAOYSA-N
Synonym 2',4'-difluoropropiophenone, 1-2,4-difluorophenyl propan-1-one, 2,4-difluoropropiophenone, 1-propanone, 1-2,4-difluorophenyl, 2,4-defluoropropiophenone, 1-2,4-difluorophenyl-1-propanone, 1-2.4-difluoro-phenyl-propan-1-one, acmc-209q3o, ksc654a2r
PubChem CID 123586
IUPAC Name 1-(2,4-difluorophenyl)propan-1-one
SMILES CCC(=O)C1=CC=C(F)C=C1F

Specifications

Color Yellow
Quantity 25 g
Formula Weight 170.16
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2′,4′-Difluoropropiophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.